Learn More
FUJIFILM Biosciences PYROSTAR ES-F TEST KIT 06ML
Catalog No. 5000450428
Encompass
endotoxin-specific kinetic turbidimetric reagent pre-dispensed for a single measurement Designed for use with the Toxinometer and to assist labs transitioning from a gel clot to a KTA methodology
Catalog No. 50-004-50428 Supplier FUJIFILM Biosciences Supplier No. WPESK0006
endotoxin-specific kinetic turbidimetric reagent pre-dispensed for a single measurement Designed for use with the Toxinometer and to assist labs transitioning from a gel clot to a KTA methodology