Learn More
Associates of Cape Cod Inc Pyrotell, 0.25 EU/mL, 5 mL/vial, 5/pk
SDP
Catalog No. NC1241126
Encompass
Pyrotell lysate was the first LAL gel-clot reagent licensed by the US FDA. It is easy to use and is available in both economical multi-test vials and convenient Single Test Vials (STV's). Pyrotell lysate is a robust reagent, producing firm, easily read clots and is resistant to interfering substances. The gel-clot test does not require sophisticated capital equipment and software and is the simplest LAL test to implement.
Catalog No. NC1241126 Supplier Associates of Cape Cod Inc Supplier No. G52505
Pyrotell lysate was the first LAL gel-clot reagent licensed by the US FDA. It is easy to use and is available in both economical multi-test vials and convenient Single Test Vials (STV's). Pyrotell lysate is a robust reagent, producing firm, easily read clots and is resistant to interfering substances. The gel-clot test does not require sophisticated capital equipment and software and is the simplest LAL test to implement.