Learn More
Associates of Cape Cod Inc Pyrotell 0.06 EU/mL, 5 vials
SDP
Catalog No. NC2030318
Encompass
Pyrotell lysate was the first LAL gel-clot reagent licensed by the US FDA. It is easy to use and is available in both economical multi-test vials and convenient Single Test Vials (STV's). Pyrotell lysate is a robust reagent, producing firm, easily read clots and is resistant to interfering substances. The gel-clot test does not require sophisticated capital equipment and software and is the simplest LAL test to implement. For standard use, a 1:1 mixture of Pyrotell lysate to sample is used. The reconstituted lysate is stable for 24 hours at 2–8°C or for up to 3 months at or below -20°C. May be frozen onceand will retain activity for as long as 3 months if stored at or below -20°C.
Catalog No. NC2030318 Supplier Associates of Cape Cod Inc Supplier No. GS0065
Pyrotell lysate was the first LAL gel-clot reagent licensed by the US FDA. It is easy to use and is available in both economical multi-test vials and convenient Single Test Vials (STV's). Pyrotell lysate is a robust reagent, producing firm, easily read clots and is resistant to interfering substances. The gel-clot test does not require sophisticated capital equipment and software and is the simplest LAL test to implement. For standard use, a 1:1 mixture of Pyrotell lysate to sample is used. The reconstituted lysate is stable for 24 hours at 2–8°C or for up to 3 months at or below -20°C. May be frozen onceand will retain activity for as long as 3 months if stored at or below -20°C.