Learn More
Associates of Cape Cod Inc PYROTELL25 VIAL/PACK
SDP
Catalog No. NC0243669
Encompass
PYROTELL GEL CLOT LAL, 0.03 EU/mL SENSITIVITY, 5mL/VIAL, 25 VIALS/PACK
Catalog No. NC0243669 Supplier Associates of Cape Cod Inc Supplier No. G500325
PYROTELL GEL CLOT LAL, 0.03 EU/mL SENSITIVITY, 5mL/VIAL, 25 VIALS/PACK