Learn More
Medchemexpress LLC HY-104065B 5mg Medchemexpress, Pyrotinib dimaleate CAS:1397922-61-0 Purity:>98%
SDP
Catalog No. 501968201
Encompass_Preferred
Medchemexpress, HY-104065B 5mg Pyrotinib dimaleate CAS:1397922-61-0 Pyrotinib dimaleate (SHR-1258 dimaleate) is a potent and selective EGFR/HER2 dual inhibitor with IC50 s of 13 and 38 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8201 Supplier Medchemexpress LLC Supplier No. HY104065B5MG
Medchemexpress, HY-104065B 5mg Pyrotinib dimaleate CAS:1397922-61-0 Pyrotinib dimaleate (SHR-1258 dimaleate) is a potent and selective EGFR/HER2 dual inhibitor with IC50 s of 13 and 38 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.