Learn More
Strem, An Ascensus Company CAS# 1257527-14-2. 100mg. 2-[(4S)-4-tert-Butyl-4,5-dihydro-2-oxazolyl]-4-(trifluoromethyl)pyridine, 98%.
SDP
Supplier:  Strem, An Ascensus Company 071050100MG
Encompass
CAS# 1257527-14-2. 100mg. 2-[(4S)-4-tert-Butyl-4,5-dihydro-2-oxazolyl]-4-(trifluoromethyl)pyridine, 98%. . Molecular Weight: 272.3. Molecular Formula: C13H15F3N2O. Color/form: off white to light yellow pwdr. Strem# 07-1050. http://www.strem.com/catalog/v/07-1050/
Catalog No. 50-206-5381