Learn More
Strem, An Ascensus Company CAS . 50mg. 2-[(4R)-4-Isobutyl-45-dihydro-2-oxazolyl]-5-(trifluoromethyl)pyridine 98% (99% ee).
SDP
Catalog No. 502110697
Encompass
50mg. 2-[(4R)-4-Isobutyl-45-dihydro-2-oxazolyl]-5-(trifluoromethyl)pyridine 98% (99% ee). . Molecular Weight 272.30. Molecular Formula C13H15F3N2O. Color/form white to yellow pwdr. Strem 07-1040. http//www.strem.com/catalog/v/07-1040/
Catalog No. 50-211-0697 Supplier Strem, An Ascensus Company Supplier No. 07104050MG
50mg. 2-[(4R)-4-Isobutyl-45-dihydro-2-oxazolyl]-5-(trifluoromethyl)pyridine 98% (99% ee). . Molecular Weight 272.30. Molecular Formula C13H15F3N2O. Color/form white to yellow pwdr. Strem 07-1040. http//www.strem.com/catalog/v/07-1040/