Learn More
Medchemexpress LLC Pyrraline | 74509-14-1 | 254.28 g/mol | C12H18N2O4 | 1 MG
SDP
Catalog No. 5000284399
Encompass_Preferred
Pyrraline is a research-grade small molecule that activates the Keap1-Nrf2 pathway and modulates NF-κB, producing antioxidant and anti-inflammatory effects useful for cellular oxidative stress and inflammation studies.
  • Activates Keap1-Nrf2 signaling and modulates NF-κB activity.
  • Produces antioxidant and anti-inflammatory responses in cellular models.
  • Upregulates antioxidant enzymes such as NQO1 and HO-1.
  • Suitable for preclinical research on oxidative stress and inflammation mechanisms.
  • Refer to the certificate of analysis for batch-specific purity and storage conditions.
Catalog No. 50-002-84399 Supplier Medchemexpress LLC Supplier No. HYW4787411MG
Pyrraline is a research-grade small molecule that activates the Keap1-Nrf2 pathway and modulates NF-κB, producing antioxidant and anti-inflammatory effects useful for cellular oxidative stress and inflammation studies.
  • Activates Keap1-Nrf2 signaling and modulates NF-κB activity.
  • Produces antioxidant and anti-inflammatory responses in cellular models.
  • Upregulates antioxidant enzymes such as NQO1 and HO-1.
  • Suitable for preclinical research on oxidative stress and inflammation mechanisms.
  • Refer to the certificate of analysis for batch-specific purity and storage conditions.