Learn More
Cayman Chemical Pyrrolidinedithiocarbamic Acid (ammonium salt), Purity: ≥98%, CAS: 5108-96-3, 10 g
SDP
Catalog No. 502984287
Encompass_Preferred
A metal chelating compound that potently and reversibly inhibits NF-κB activation (100 µM); prevents TNF-α gene expression and protein production (300 µM); arrests the cell cycle at G1; has antioxidant properties
Catalog No. 50-298-4287 Supplier Cayman Chemical Supplier No. 2071310 G
A metal chelating compound that potently and reversibly inhibits NF-κB activation (100 µM); prevents TNF-α gene expression and protein production (300 µM); arrests the cell cycle at G1; has antioxidant properties