Learn More
Enzo Life Sciences Pyrrolidinedithiocarbamic acid. ammonium salt (5g). CAS: 5108-96-3
SDP
Supplier:  Enzo Life Sciences ALX400002G005
Encompass_Preferred
Inhibitor of NF-κB activation (induced by either PMA or TNF-α) in various cell types. Alternative name: Ammoniumpyrrolidinedithiocarbamate, Ammoniumtetramethylenedithiocarbamate. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9065