Learn More
Medchemexpress LLC Pyrrole-2-carboxaldehyde | 1003-29-8 | 99.99% | C₅H₅NO | 1 G
SDP
Catalog No. 5000434830
Encompass_Preferred
Pyrrole-2-carboxaldehyde (2-Formylpyrrole) is a five-membered N-heterocyclic system that exhibits distinct vibrational and electronic characteristics. It is observed in a dimeric form in the solid phase and a monomeric form in solution.
  • Five-membered N-heterocyclic system
  • Pyrrole group acts as a proton donor
  • Carbonyl group functions as a proton acceptor
  • Exhibits vibrational and electronic characteristics
  • Exists as a dimeric form in the solid phase
  • Exists as a monomeric form in solution
Catalog No. 50-004-34830 Supplier Medchemexpress LLC Supplier No. HY778171G
Pyrrole-2-carboxaldehyde (2-Formylpyrrole) is a five-membered N-heterocyclic system that exhibits distinct vibrational and electronic characteristics. It is observed in a dimeric form in the solid phase and a monomeric form in solution.
  • Five-membered N-heterocyclic system
  • Pyrrole group acts as a proton donor
  • Carbonyl group functions as a proton acceptor
  • Exhibits vibrational and electronic characteristics
  • Exists as a dimeric form in the solid phase
  • Exists as a monomeric form in solution