Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrole-2-carboxaldehyde | 1003-29-8 | 99.99% | C5H5NO | 25 G
SDP

Catalog No. 5000358524
Encompass_Preferred

Pyrrole-2-carboxaldehyde (2-Formylpyrrole) is a five-membered N-heterocyclic system, acting as both a proton donor and acceptor. It displays unique vibrational and electronic characteristics, existing as a dimer in solid phase and a monomer in solution.

  • Exhibits vibrational and electronic characteristics
  • Exists as dimer in solid phase, monomer in solution
  • Five-membered N-heterocyclic system
  • Functions as proton donor and acceptor
  • Solid, white to light brown appearance
  • Store at 4°C, protected from light, under nitrogen
  • Store in solvent: -80°C (6 months), -20°C (1 month)
  • Soluble in DMSO (100 mg/mL) with sonication
  • Suitable for various in vivo applications

Catalog No. 50-003-58524 Supplier Medchemexpress LLC Supplier No. 25GHY77817
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrrole-2-carboxaldehyde (2-Formylpyrrole) is a five-membered N-heterocyclic system, acting as both a proton donor and acceptor. It displays unique vibrational and electronic characteristics, existing as a dimer in solid phase and a monomer in solution.

  • Exhibits vibrational and electronic characteristics
  • Exists as dimer in solid phase, monomer in solution
  • Five-membered N-heterocyclic system
  • Functions as proton donor and acceptor
  • Solid, white to light brown appearance
  • Store at 4°C, protected from light, under nitrogen
  • Store in solvent: -80°C (6 months), -20°C (1 month)
  • Soluble in DMSO (100 mg/mL) with sonication
  • Suitable for various in vivo applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.