Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrole-2-carboxaldehyde | 1003-29-8 | >99.99% | C5H5NO | 5 G
SDP

Catalog No. 5000433468
Encompass_Preferred

Pyrrole-2-carboxaldehyde is a five-membered N-heterocyclic system containing a pyrrole group as a proton donor and a carbonyl group as a proton acceptor. It exhibits vibrational and electronic characteristics that establish its dimeric form in solid phase and monomeric form in solution phase.

  • Solid appearance
  • White to light brown color
  • Molecular weight of 95.10
  • Classified as pyrrole alkaloids
  • Initial source from plants
  • Store at 4°C, protected from light and under nitrogen
  • Soluble in DMSO at 100 mg/mL (1051.52 mM)

Catalog No. 50-004-33468 Supplier Medchemexpress LLC Supplier No. HY778175G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrrole-2-carboxaldehyde is a five-membered N-heterocyclic system containing a pyrrole group as a proton donor and a carbonyl group as a proton acceptor. It exhibits vibrational and electronic characteristics that establish its dimeric form in solid phase and monomeric form in solution phase.

  • Solid appearance
  • White to light brown color
  • Molecular weight of 95.10
  • Classified as pyrrole alkaloids
  • Initial source from plants
  • Store at 4°C, protected from light and under nitrogen
  • Soluble in DMSO at 100 mg/mL (1051.52 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.