Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrole-2-carboxaldehyde | 1003-29-8 | MFCD00005217 | 99.99% | C5H5NO | 10 G
SDP

Catalog No. 5000435025
Encompass_Preferred

Pyrrole-2-carboxaldehyde is a five-membered N-heterocyclic system featuring a pyrrole group as a proton donor and a carbonyl group as a proton acceptor. It exists in a dimeric form in the solid phase and a monomeric form in solution, as indicated by its vibrational and electronic characteristics.

  • High purity of 99.99%
  • Molecular weight: 95.10
  • Chemical formula: C5H5NO
  • Appears as a solid
  • Color ranges from white to light brown
  • Classified as alkaloids and pyrrole alkaloids
  • Initial source is plants other families
  • Store at 4°C, protected from light and under nitrogen

Catalog No. 50-004-35025 Supplier Medchemexpress LLC Supplier No. HY7781710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrrole-2-carboxaldehyde is a five-membered N-heterocyclic system featuring a pyrrole group as a proton donor and a carbonyl group as a proton acceptor. It exists in a dimeric form in the solid phase and a monomeric form in solution, as indicated by its vibrational and electronic characteristics.

  • High purity of 99.99%
  • Molecular weight: 95.10
  • Chemical formula: C5H5NO
  • Appears as a solid
  • Color ranges from white to light brown
  • Classified as alkaloids and pyrrole alkaloids
  • Initial source is plants other families
  • Store at 4°C, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.