Learn More
Selleck Chemical LLC Pyrrole-2-carboxylic acid
SDP
Catalog No. 5000002889
Encompass_Preferred
Pyrrole-2-carboxylic acid (2-Pyrrolecarboxylic acid) is a degradation product of sialic acids and a derivative of the oxidation of the D-hydroxyproline isomers by mammalian D-amino acid oxidase
Catalog No. 50-000-02889 Supplier Selleck Chemical LLC Supplier No. S583225MG
Pyrrole-2-carboxylic acid (2-Pyrrolecarboxylic acid) is a degradation product of sialic acids and a derivative of the oxidation of the D-hydroxyproline isomers by mammalian D-amino acid oxidase