Learn More
Medchemexpress LLC Pyrrole-2-carboxylic acid | 634-97-9 | 111.10 | 1 ML
SDP
Catalog No. 5000447328
Encompass_Preferred
Pyrrole-2-carboxylic acid is a natural alkaloid originating from the marine bacterium Pelomonas puraquae sp. Nov. This biological form of pyrrole is frequently found in natural compounds and serves as a precursor in biosynthetic pathways for various pyrrole derivatives, including clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin. This product is supplied as a 10 mM solution in DMSO.
  • Natural alkaloid derived from marine bacterium
  • Precursor in biosynthetic pathways
  • Used in the synthesis of various pyrrole derivatives
Catalog No. 50-004-47328 Supplier Medchemexpress LLC Supplier No. HYW00196310MM/1ML
Pyrrole-2-carboxylic acid is a natural alkaloid originating from the marine bacterium Pelomonas puraquae sp. Nov. This biological form of pyrrole is frequently found in natural compounds and serves as a precursor in biosynthetic pathways for various pyrrole derivatives, including clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin. This product is supplied as a 10 mM solution in DMSO.
  • Natural alkaloid derived from marine bacterium
  • Precursor in biosynthetic pathways
  • Used in the synthesis of various pyrrole derivatives