Learn More
Medchemexpress LLC Pyrrole-2-carboxylic acid | 634-97-9 | 111.10 | 25 G
SDP
Catalog No. 5000450095
Encompass_Preferred
Pyrrole-2-carboxylic acid is a natural alkaloid found in the marine bacterium *Pelomonas puraquae* sp. Nov. It is a biological form of pyrrole frequently encountered in natural compounds and acts as a precursor for various pyrrole derivatives in biosynthetic processes, including clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin.
  • Natural alkaloid sourced from marine bacterium *Pelomonas puraquae* sp. Nov.
  • Acts as a precursor for various pyrrole derivatives.
  • Exhibits antineuroinflammatory activity in LPS-induced mouse BV-2 cells.
Catalog No. 50-004-50095 Supplier Medchemexpress LLC Supplier No. 25GHYW001963
Pyrrole-2-carboxylic acid is a natural alkaloid found in the marine bacterium *Pelomonas puraquae* sp. Nov. It is a biological form of pyrrole frequently encountered in natural compounds and acts as a precursor for various pyrrole derivatives in biosynthetic processes, including clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin.
  • Natural alkaloid sourced from marine bacterium *Pelomonas puraquae* sp. Nov.
  • Acts as a precursor for various pyrrole derivatives.
  • Exhibits antineuroinflammatory activity in LPS-induced mouse BV-2 cells.