Learn More
Medchemexpress LLC Pyrrole-2-carboxylic acid | 634-97-9 | 111.10 | 5 G
SDP
Catalog No. 5000447330
Encompass_Preferred
Pyrrole-2-carboxylic acid is a natural alkaloid found in the marine bacterium *Pelomonas puraquae* sp. Nov. This biological form of pyrrole is frequently encountered in natural compounds.
  • Acts as a precursor for various pyrrole derivatives
  • Involved in biosynthetic pathways for clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin
Catalog No. 50-004-47330 Supplier Medchemexpress LLC Supplier No. HYW0019635G
Pyrrole-2-carboxylic acid is a natural alkaloid found in the marine bacterium *Pelomonas puraquae* sp. Nov. This biological form of pyrrole is frequently encountered in natural compounds.
  • Acts as a precursor for various pyrrole derivatives
  • Involved in biosynthetic pathways for clorobiocin, coumermycin A1, pyoluteorin, leupyrrins, and undecylprodigiosin