Learn More
Apexbio Technology LLC Pyrrole-2-carboxylic acid 634-97-9 500mg
SDP
Catalog No. 5000094660
Encompass_Preferred
Pyrrole-2-carboxylic acid (CAS 634-97-9) is a heterocyclic organic compound characterized by a carboxyl substituent at the 2-position of the pyrrole ring This molecule has been investigated in biomedical research as an inhibitor of certain enzymatic pathways involving pyrrole derivatives including roles in microbial metabolism and secondary metabolite biosynthesis Pyrrole-2-carboxylic acid is utilized in studies examining enzyme specificity metabolic pathway modulation and as a precursor in the synthesis of pharmacologically relevant molecules Its defined structure makes it a valuable tool for probing pyrrole-related biochemical mechanisms
Catalog No. 50-000-94660 Supplier Apexbio Technology LLC Supplier No. C7008500
Pyrrole-2-carboxylic acid (CAS 634-97-9) is a heterocyclic organic compound characterized by a carboxyl substituent at the 2-position of the pyrrole ring This molecule has been investigated in biomedical research as an inhibitor of certain enzymatic pathways involving pyrrole derivatives including roles in microbial metabolism and secondary metabolite biosynthesis Pyrrole-2-carboxylic acid is utilized in studies examining enzyme specificity metabolic pathway modulation and as a precursor in the synthesis of pharmacologically relevant molecules Its defined structure makes it a valuable tool for probing pyrrole-related biochemical mechanisms