Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyrrole

Catalog No. 13170925L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-170-925L 2.5 L
13-170-9100ML 100 mL
13-170-925ML 25 mL
13-170-9500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-170-925L Supplier Sigma Aldrich Supplier No. 1317092.5L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pyrrole

Specifications

CAS 109-97-7
Melting Point -23°C (lit.)
Density 0.967 g/mL (at 25°C (literature))
Boiling Point 131°C (lit.)
Molecular Formula C4H5N
Refractive Index n20/D 1.508 (literature)
Linear Formula C4H5N
MDL Number MFCD00005216
Quantity 2.5 L
Synonym Azole; Divinylenimine; Imidole
Molecular Weight (g/mol) 67.09
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.