Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrole, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019682 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9682 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9682 Supplier MilliporeSigma Supelco Supplier No. 498171ML
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 109-97-7
Vapor Density 2.31 (vs. air)
Density 0.967 g/mL (at 25°C)
Flash Point 36°C - Closed Cup
Physical Form Neat
Linear Formula C4H5 N
MDL Number MFCD00005216
Melting Point -23°C
Boiling Point 131°C
Synonym Azole; Divinylenimine; Imidole
Type Analytical Standard
Quantity 1 mL
Formula Weight 67.09
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.