Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyrrole 98 fCC fG1KG
Catalog No. 502902396
Encompass
Pyrrole is one of the flavor compounds that is formed in thermally processed foods due to the Maillard reaction.
Catalog No. 50-290-2396 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3386051KG
Pyrrole is one of the flavor compounds that is formed in thermally processed foods due to the Maillard reaction.