Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrole, 99%, extra pure

Catalog No. ac157710250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157710250 25 g Glass bottle
AC157711000 100 g Glass bottle
AC157715000 500 g Glass bottle
AC157710025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC157710250 Supplier Thermo Scientific Chemicals Supplier No. 157710250
Add to Cart

CAS: 109-97-7 | C4H5N | 67.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-97-7
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00005216
InChI Key KAESVJOAVNADME-UHFFFAOYSA-N
Synonym pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
PubChem CID 8027
ChEBI CHEBI:19203
IUPAC Name 1H-pyrrole
SMILES N1C=CC=C1

Specifications

Melting Point -23.0°C
Color Brown to Colorless
Density 0.9600g/mL
Boiling Point 129.0°C to 131.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5080 to 1.5100
Quantity 25 g
Beilstein 20,159
Merck Index 15,8128
Specific Gravity 0.96
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: insoluble in dilute alkalies, soluble in alcohol, ether and dilute acids, soluble in most organic solvents
Viscosity 1.317 mPa.s (20°C)
Formula Weight 67.09
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Pyrrole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes serious eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 203-724-7
RTECSNumber UX9275000
TSCA TSCA
Recommended Storage Darkens on storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.