Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrole 99.0+%, TCI America™
SDP

Catalog No. P0574100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0574100ML 100 mL
P057425ML 25 mL
P0574500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0574100ML Supplier TCI America Supplier No. P0574100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109-97-7
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00005216
InChI Key KAESVJOAVNADME-UHFFFAOYSA-N
Synonym pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
PubChem CID 8027
ChEBI CHEBI:19203
IUPAC Name 1H-pyrrole
SMILES N1C=CC=C1

Specifications

Melting Point -23°C
Color Yellow
Boiling Point 130°C
Quantity 100 mL
UN Number 1992
Formula Weight 67.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyrrole
EINECSNumber (5)-0100
RTECSNumber UX9275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.