Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrolidin-3-ol-d5 | 1219805-52-3 | 92.15 g/mol | C4H4D5NO | 10 MG
SDP

Catalog No. 5000209638
Encompass_Preferred

Pyrrolidin-3-ol-d5 is the deuterium-labeled analogue of pyrrolidin-3-ol used as a stable isotope tracer and internal standard in analytical and research applications.

  • Deuterated tracer for mass spectrometry and isotope dilution assays.
  • Isotopic enrichment: 97 atom % D.
  • Molecular formula C4H4D5NO.
  • Molecular weight 92.15 g/mol.
  • Available in small quantities suitable for analytical standards, e.g., 10 mg packaging.

Catalog No. 50-002-09638 Supplier Medchemexpress LLC Supplier No. HYW042206S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrrolidin-3-ol-d5 is the deuterium-labeled analogue of pyrrolidin-3-ol used as a stable isotope tracer and internal standard in analytical and research applications.

  • Deuterated tracer for mass spectrometry and isotope dilution assays.
  • Isotopic enrichment: 97 atom % D.
  • Molecular formula C4H4D5NO.
  • Molecular weight 92.15 g/mol.
  • Available in small quantities suitable for analytical standards, e.g., 10 mg packaging.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.