Learn More
Medchemexpress LLC Pyrrolidin-3-ol-d5 | 1219805-52-3 | 92.15 g/mol | C4H4D5NO | 5 MG
SDP
Catalog No. 5000209259
Encompass_Preferred
Pyrrolidin-3-ol-d5 is a deuterium-labeled analogue of pyrrolidin-3-ol supplied as a small milligram vial for use as a tracer and internal standard in quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled tracer and internal standard.
  • Used for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Offered in small milligram pack sizes for analytical workflows.
  • Storage: pure form -20°C; in solvent -80°C (short term) or -20°C.
Catalog No. 50-002-09259 Supplier Medchemexpress LLC Supplier No. HYW042206S5MG
Pyrrolidin-3-ol-d5 is a deuterium-labeled analogue of pyrrolidin-3-ol supplied as a small milligram vial for use as a tracer and internal standard in quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled tracer and internal standard.
  • Used for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Offered in small milligram pack sizes for analytical workflows.
  • Storage: pure form -20°C; in solvent -80°C (short term) or -20°C.