Learn More
Medchemexpress LLC Pyrrolidine, 3-(2,3-difluorophenyl)-3-methoxy-, (3S)-, (2E)-2-butenedioate (1:1) | 2251806-70-7 | 98.3% | 329.30 | 25 MG
SDP
Catalog No. 5000351794
Encompass_Preferred
Pirepemat fumarate (IRL752 fumarate) is a cortical-preferring catecholamine- and cognition-promoting agent. It is used for the study of Parkinson's disease and shows high affinities for 5-HT and NA-related targets. It can reverse deficits resulting from hypomonoaminergic function.
  • Cortical-preferring catecholamine- and cognition-promoting agent
  • Used for the study of Parkinson's disease
  • Shows highest in vitro affinities for 5-HT and NA-related targets
  • Reverses deficits from hypomonoaminergic function
  • White to off-white solid appearance
  • Store at 4°C, sealed and away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture
Catalog No. 50-003-51794 Supplier Medchemexpress LLC Supplier No. 25MGHY137447A
Pirepemat fumarate (IRL752 fumarate) is a cortical-preferring catecholamine- and cognition-promoting agent. It is used for the study of Parkinson's disease and shows high affinities for 5-HT and NA-related targets. It can reverse deficits resulting from hypomonoaminergic function.
  • Cortical-preferring catecholamine- and cognition-promoting agent
  • Used for the study of Parkinson's disease
  • Shows highest in vitro affinities for 5-HT and NA-related targets
  • Reverses deficits from hypomonoaminergic function
  • White to off-white solid appearance
  • Store at 4°C, sealed and away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture