Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyrrolidine

Catalog No. 394238100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-423-8100ML 100 mL
39-423-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-423-8100ML Supplier Sigma Aldrich Supplier No. 394238100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Pyrrolidine is a five membered cyclic amine. it is a nitrogen heterocycle that forms the nucleus of various bioactive molecules. it can be synthesized by reacting 1,4-diaminobutanes with acids. it behaves as a strong base in employed in certain homogeneous nonaqueous reactions. its crystalline structure has been investigated. its reaction on the ge(100)-2x1 surface and si(100)-2x1 surface has been investigated. pyrrolidine has been found to be one of the component of volatile amine fraction in the brewing process. a study on the kinetics of pyrrolidinolysis of phthalimide (pth) shows second- and third-order terms in the rate law. biodegradation of pyrrolidine by mycobacterium strains has been studied.pyrrolidine may be used in the following studies:
  • Synthesis of pyrrolidine derivatives of pyrimidine
  • As a catalyst in the selective monoallylation of allylic alcohols
  • To capture carbondioxide in a bubble column reactor using pyrrolidine aqueous solutions by gas-liquid absorption

Specifications

CAS 123-75-1
Density 0.852 g/mL (at 25°C (literature))
Boiling Point 87°C to 88°C (760 mmHg)
Molecular Formula C4H9N
Refractive Index n20/D 1.443 (literature)
Linear Formula C4H9N
MDL Number MFCD00005249
Quantity 100 mL
Synonym Tetrahydropyrrole; Tetramethyleneimine
Molecular Weight (g/mol) 71.12
Percent Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.