Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyrrolidine

Catalog No. P73803100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P73803100ML 100 mL
P73803500ML 500 mL
P738035ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P73803100ML Supplier Sigma Aldrich Supplier No. P73803100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Pyrrolidine has been used for the synthesis of n-benzoyl pyrrolidine from benzaldehyde via oxidative amination. it may be used as a catalyst for the synthesis of n-sulfinyl aldimines from carbonyl compounds and sulfonamides.pyrrolidine can also be used to synthesize:
  • Taddol-pyrrolidine phosphoramidite, a ligand for rhodium-catalyzed [2+2+2] cycloaddition of pentenyl isocyanate and 4- ethynylanisole
  • H,4 pyrrolidinequin-bam (αpbamα), a selective catalyst for the aza-henry addition of nitroalkanes to aryl aldimines.{88]
  • 1,2,3,3a,4,9-hexahydropyrrolo[2,1-b]quinazoline by reacting with o-aminobenzaldehyde

Specifications

CAS 123-75-1
Density 0.852 g/mL (at 25°C (literature))
Boiling Point 87°C to 88°C (760 mmHg)
Molecular Formula C4H9N
Refractive Index n20/D 1.443 (literature)
Linear Formula C4H9N
MDL Number MFCD00005249
Quantity 100 mL
Synonym Tetrahydropyrrole; Tetramethyleneimine
Molecular Weight (g/mol) 71.12
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.