Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrolidine, TCI America™
SDP

Catalog No. P057625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P057625ML 25 mL
P0576500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P057625ML Supplier TCI America Supplier No. P057625ML

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD00005249
InChI Key RWRDLPDLKQPQOW-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name pyrrolidine
SMILES C1CCNC1

Specifications

Melting Point -63°C
Color Colorless
Boiling Point 88°C
Quantity 25 mL
UN Number 1922
Formula Weight 71.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyrrolidine
EINECSNumber (5)-0103
RTECSNumber UX9650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.