Learn More
Medchemexpress LLC HY-18738 100mg Medchemexpress, Pyrrolidinedithiocarbamate (ammonium) CAS:5108-96-3 Purity:>98%
SDP
Catalog No. 502082080
Encompass_Preferred
Medchemexpress, HY-18738 100mg Pyrrolidinedithiocarbamate (ammonium) CAS:5108-96-3 Pyrrolidinedithiocarbamate ammonium (Ammonium pyrrolidinedithiocarbamate) is a selective NF-κB inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2080 Supplier Medchemexpress LLC Supplier No. HY18738100MG
Medchemexpress, HY-18738 100mg Pyrrolidinedithiocarbamate (ammonium) CAS:5108-96-3 Pyrrolidinedithiocarbamate ammonium (Ammonium pyrrolidinedithiocarbamate) is a selective NF-κB inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.