Learn More
Cayman Chemical PyrrolIdIno PAF C-16 10mg
SDP
Catalog No. 502893951
Encompass_Preferred
A synthetic analog of PAF C-16 with approximately 3-10 times increased potency
Catalog No. 50-289-3951 Supplier Cayman Chemical Supplier No. 6090910 MG
A synthetic analog of PAF C-16 with approximately 3-10 times increased potency