Learn More
Medchemexpress LLC Pyrroline-5-carboxylate (sodium) | 72978-16-6 | 98.0% | 135.10 | 25 MG
SDP
Catalog No. 5000348543
Encompass_Preferred
Pyrroline-5-carboxylate sodium is involved in amino acid metabolism within organisms, serving as an intermediate product in both the biosynthesis and catabolism of proline. It plays a crucial role during pathogen infection and in response to abiotic stress. This product is strictly for research use only.
  • Participates in amino acid metabolism
  • Intermediate product of proline biosynthesis and catabolism
  • Plays a role during pathogen infection and abiotic stress
  • Suitable for laboratory chemical applications
  • Used in the manufacture of substances
Catalog No. 50-003-48543 Supplier Medchemexpress LLC Supplier No. 25MGHY138354A
Pyrroline-5-carboxylate sodium is involved in amino acid metabolism within organisms, serving as an intermediate product in both the biosynthesis and catabolism of proline. It plays a crucial role during pathogen infection and in response to abiotic stress. This product is strictly for research use only.
  • Participates in amino acid metabolism
  • Intermediate product of proline biosynthesis and catabolism
  • Plays a role during pathogen infection and abiotic stress
  • Suitable for laboratory chemical applications
  • Used in the manufacture of substances