Learn More
Medchemexpress LLC Pyrroline-5-carboxylate sodium | 72978-16-6 | 98.03% | 135.10 | 100 MG
SDP
Catalog No. 5000350460
Encompass_Preferred
Pyrroline-5-carboxylate sodium is an intermediate product involved in amino acid metabolism, specifically in the biosynthesis and catabolism of proline within organisms. It also plays a role in plant defense mechanisms against invading pathogens and abiotic stress. This product is intended for research use only.
  • Participates in amino acid metabolism
  • Intermediate product of proline biosynthesis and catabolism
  • Involved during pathogen infection
  • Involved during abiotic stress
  • Intended for research use only
Catalog No. 50-003-50460 Supplier Medchemexpress LLC Supplier No. HY138354A100MG
Pyrroline-5-carboxylate sodium is an intermediate product involved in amino acid metabolism, specifically in the biosynthesis and catabolism of proline within organisms. It also plays a role in plant defense mechanisms against invading pathogens and abiotic stress. This product is intended for research use only.
  • Participates in amino acid metabolism
  • Intermediate product of proline biosynthesis and catabolism
  • Involved during pathogen infection
  • Involved during abiotic stress
  • Intended for research use only