Learn More
Medchemexpress LLC Pyrroline-5-carboxylate sodium | 72978-16-6 | 99.8% | 135.10 g·mol-1 | C5H6NNaO2 | 5 MG
SDP
Catalog No. 5000222905
Encompass_Preferred
Pyrroline-5-carboxylate sodium is the sodium salt of pyrroline-5-carboxylic acid supplied as a white to off-white solid for research use. It functions as an intermediate in proline biosynthesis and catabolism and is provided in small pre-weighed packages for biochemical and synthetic applications. A certificate of analysis is available.
Catalog No. 50-002-22905 Supplier Medchemexpress LLC Supplier No. HY138354A5MG
Pyrroline-5-carboxylate sodium is the sodium salt of pyrroline-5-carboxylic acid supplied as a white to off-white solid for research use. It functions as an intermediate in proline biosynthesis and catabolism and is provided in small pre-weighed packages for biochemical and synthetic applications. A certificate of analysis is available.