Learn More
Medchemexpress LLC Pyrroline-5-carboxylate sodium | 72978-16-6 | 99.8% | C5H6NNaO2 | 10MG
SDP
Catalog No. 5000262013
Encompass_Preferred
Pyrroline-5-carboxylate sodium is the sodium salt form of pyrroline-5-carboxylate, an intermediate in proline biosynthesis and catabolism. Supplied for research use, it is intended as a biochemical intermediate and reagent for metabolic and enzymatic studies. Store sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • High purity: 99.79% reported
  • Available in small research pack sizes (5-500 mg)
  • Molecular weight 135.10 and formula C5H6NNaO2
  • Suitable for metabolic and enzymatic assays
  • Recommended sealed storage to avoid moisture
Catalog No. 50-002-62013 Supplier Medchemexpress LLC Supplier No. HY138354A10MG
Pyrroline-5-carboxylate sodium is the sodium salt form of pyrroline-5-carboxylate, an intermediate in proline biosynthesis and catabolism. Supplied for research use, it is intended as a biochemical intermediate and reagent for metabolic and enzymatic studies. Store sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • High purity: 99.79% reported
  • Available in small research pack sizes (5-500 mg)
  • Molecular weight 135.10 and formula C5H6NNaO2
  • Suitable for metabolic and enzymatic assays
  • Recommended sealed storage to avoid moisture