Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrolnitrin | 1018-71-9 | MFCD00865605 | 99.8% | 257.07 g/mol | C10H6Cl2N2O2 | 5 MG
SDP

Catalog No. 5000262223
Encompass_Preferred

Pyrrolnitrin is a chlorinated pyrrole antibiotic small molecule used as a research reagent with activity against fungi, yeast, and gram-positive bacteria. Supplied as a high-purity powder for laboratory use; for research use only - not for human or veterinary use.

  • High purity: 99.8%.
  • Molecular weight: 257.07 g/mol.
  • Chemical formula: C10H6Cl2N2O2.
  • Available in 5 mg vials.
  • Powder stability: -20°C for up to 3 years.
  • Suitable for antifungal and gram-positive antibacterial research assays.

Catalog No. 50-002-62223 Supplier Medchemexpress LLC Supplier No. HY1337045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrrolnitrin is a chlorinated pyrrole antibiotic small molecule used as a research reagent with activity against fungi, yeast, and gram-positive bacteria. Supplied as a high-purity powder for laboratory use; for research use only - not for human or veterinary use.

  • High purity: 99.8%.
  • Molecular weight: 257.07 g/mol.
  • Chemical formula: C10H6Cl2N2O2.
  • Available in 5 mg vials.
  • Powder stability: -20°C for up to 3 years.
  • Suitable for antifungal and gram-positive antibacterial research assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.