Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrolo[2,1-b][1,3]oxazepine-7-carboxamide, octahydro-8,8-dimethyl-4-[[(2S)-2-(methylamino)-1-oxopropyl]ami | 1422180-49-1 | MFCD34469389 | 98.4% | 456.58 g/mol | C25H36N4O4 | 5MG
SDP

Catalog No. 5000235084
Encompass_Preferred

SBP-0636457 is a small-molecule SMAC mimetic and inhibitor of apoptosis protein (IAP) antagonist used for preclinical cancer research. It binds the BIR domains of IAP proteins (Ki = 0.27 μM) and has shown TRAIL-sensitizing activity in cell models, supporting use in both in vitro and in vivo studies.

  • Binds inhibitor of apoptosis proteins with Ki of 0.27 μM.
  • Demonstrated TRAIL-sensitizing activity (EC50 9 nM in MDA-MB-231 cells).
  • High purity suitable for biological assays.
  • Soluble in DMSO (≈45.7 mg/mL) and compatible with several in vivo formulations.
  • Storage recommendations available for solid and solution forms to preserve stability.
  • No cytotoxicity observed in selected breast cancer cell lines up to 20 μM.

Catalog No. 50-002-35084 Supplier Medchemexpress LLC Supplier No. HY1253785MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SBP-0636457 is a small-molecule SMAC mimetic and inhibitor of apoptosis protein (IAP) antagonist used for preclinical cancer research. It binds the BIR domains of IAP proteins (Ki = 0.27 μM) and has shown TRAIL-sensitizing activity in cell models, supporting use in both in vitro and in vivo studies.

  • Binds inhibitor of apoptosis proteins with Ki of 0.27 μM.
  • Demonstrated TRAIL-sensitizing activity (EC50 9 nM in MDA-MB-231 cells).
  • High purity suitable for biological assays.
  • Soluble in DMSO (≈45.7 mg/mL) and compatible with several in vivo formulations.
  • Storage recommendations available for solid and solution forms to preserve stability.
  • No cytotoxicity observed in selected breast cancer cell lines up to 20 μM.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.