Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrolo[3,4-b]pyrrole-5(1H)-carboxamide, N-[4-[(3-ethynylphenyl)amino]-7-methoxy-6-quinazolinyl] | 1353644-70-8 | 99.7% | 442.51 g/mol | C25H26N6O2 | 25 MG
SDP

Catalog No. 5000243800
Encompass_Preferred

Theliatinib is a potent, ATP-competitive, orally active, and highly selective epidermal growth factor receptor (EGFR) inhibitor supplied as a research chemical for biochemical and cellular studies. It shows low-nanomolar potency against wild-type EGFR and measurable activity against clinically relevant mutant forms.

  • Potent, ATP-competitive EGFR inhibitor with Ki = 0.05 nM and IC50 = 3 nM.
  • Active against EGFR T790M/L858R mutant (IC50 ≈ 22 nM).
  • High purity (~99.7%) suitable for biochemical and cellular assays.
  • Molecular weight 442.51 g/mol; formula C25H26N6O2.
  • Soluble in DMSO and supplied for laboratory research use only.

Catalog No. 50-002-43800 Supplier Medchemexpress LLC Supplier No. HY10406625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Theliatinib is a potent, ATP-competitive, orally active, and highly selective epidermal growth factor receptor (EGFR) inhibitor supplied as a research chemical for biochemical and cellular studies. It shows low-nanomolar potency against wild-type EGFR and measurable activity against clinically relevant mutant forms.

  • Potent, ATP-competitive EGFR inhibitor with Ki = 0.05 nM and IC50 = 3 nM.
  • Active against EGFR T790M/L858R mutant (IC50 ≈ 22 nM).
  • High purity (~99.7%) suitable for biochemical and cellular assays.
  • Molecular weight 442.51 g/mol; formula C25H26N6O2.
  • Soluble in DMSO and supplied for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.