Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrrolo[3,4-b]pyrrole-5(1H)-carboxamide, N-[4-[(3-ethynylphenyl)amino]-7-methoxy-6-quinazolinyl] | 1353644-70-8 | 99.7% | 442.51 g/mol | C25H26N6O2 | 50 MG
SDP

Catalog No. 5000243817
Encompass_Preferred

Theliatinib is a potent, ATP-competitive, orally active, and highly selective epidermal growth factor receptor (EGFR) inhibitor provided for research use in preclinical biochemical and cell-based studies.

  • Potent ATP-competitive EGFR inhibitor with low-nanomolar activity.
  • High chemical purity (99.72%).
  • Molecular weight 442.51 g/mol; formula C25H26N6O2.
  • Soluble in DMSO (~1 mg/mL) with recommended warming and pH adjustment.
  • Stable when stored under recommended powder and solution conditions.
  • Suitable for studies of wild-type and mutant EGFR signaling.

Catalog No. 50-002-43817 Supplier Medchemexpress LLC Supplier No. HY10406650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Theliatinib is a potent, ATP-competitive, orally active, and highly selective epidermal growth factor receptor (EGFR) inhibitor provided for research use in preclinical biochemical and cell-based studies.

  • Potent ATP-competitive EGFR inhibitor with low-nanomolar activity.
  • High chemical purity (99.72%).
  • Molecular weight 442.51 g/mol; formula C25H26N6O2.
  • Soluble in DMSO (~1 mg/mL) with recommended warming and pH adjustment.
  • Stable when stored under recommended powder and solution conditions.
  • Suitable for studies of wild-type and mutant EGFR signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.