Learn More
Medchemexpress LLC Pyrroloquinoline quinone-13C3 sodium | 72909-34-3 | 99.3% | 421.11 g·mol⁻¹ | C1113C3H2N2Na4O8 | 1 MG
SDP
Catalog No. 5000215872
Encompass_Preferred
Pyrroloquinoline quinone-13C3 sodium is the 13C3 isotopically labeled sodium salt of pyrroloquinoline quinone, supplied as a high-purity tracer and internal standard for quantitative analytical methods. It is intended for research use in studies of redox chemistry and biological activity, and for method development and validation in mass spectrometry and NMR-based assays.
  • Isotopically labeled sodium salt for tracer and internal standard use.
  • High purity suitable for quantitative analytical methods.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Supports studies of redox activity and biological function.
  • Supplied in small, weighed quantities for research applications.
Catalog No. 50-002-15872 Supplier Medchemexpress LLC Supplier No. HY100196S11MG
Pyrroloquinoline quinone-13C3 sodium is the 13C3 isotopically labeled sodium salt of pyrroloquinoline quinone, supplied as a high-purity tracer and internal standard for quantitative analytical methods. It is intended for research use in studies of redox chemistry and biological activity, and for method development and validation in mass spectrometry and NMR-based assays.
  • Isotopically labeled sodium salt for tracer and internal standard use.
  • High purity suitable for quantitative analytical methods.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Supports studies of redox activity and biological function.
  • Supplied in small, weighed quantities for research applications.