Learn More
Medchemexpress LLC HY-100196 5mg Medchemexpress, Pyrroloquinoline quinone CAS:72909-34-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1001965MG
Encompass_Preferred
Medchemexpress, HY-100196 5mg Pyrroloquinoline quinone CAS:72909-34-3 Pyrroloquinoline quinone (PQQ), a redox co-factor, is an anionic, redox-cycling orthoquinone. Pyrroloquinoline quinone is isolated from cultures of methylotropic bacteria and tissues of mammals. Pyrroloquinoline quinone is an essential nutrient for mammals and is important for immune function[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9447