Learn More
Medchemexpress LLC Pyrroloquinoline quinone disodium salt | 122628-50-6 | 98.0% | 374.17 g/mol | C14H4N2Na2O8 | 10 MG
SDP
Catalog No. 5000215830
Encompass_Preferred
Pyrroloquinoline quinone disodium salt is a water-soluble redox cofactor used as a research reagent in studies of cellular metabolism, mitochondrial function, and oxidative stress. The disodium salt form enhances solubility for aqueous assays and small-scale formulations, and is typically supplied with high purity for experimental reproducibility.
  • Water-soluble form suitable for aqueous assays.
  • Acts as a redox cofactor for metabolism and mitochondrial studies.
  • High purity supports reproducible experimental results.
  • Appropriate for biochemical and cell biology applications.
  • Available in small quantities for screening and pilot studies.
Catalog No. 50-002-15830 Supplier Medchemexpress LLC Supplier No. HY100196A10MG
Pyrroloquinoline quinone disodium salt is a water-soluble redox cofactor used as a research reagent in studies of cellular metabolism, mitochondrial function, and oxidative stress. The disodium salt form enhances solubility for aqueous assays and small-scale formulations, and is typically supplied with high purity for experimental reproducibility.
  • Water-soluble form suitable for aqueous assays.
  • Acts as a redox cofactor for metabolism and mitochondrial studies.
  • High purity supports reproducible experimental results.
  • Appropriate for biochemical and cell biology applications.
  • Available in small quantities for screening and pilot studies.