Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrroloquinoline quinone disodium salt | 122628-50-6 | 98.0% | 374.17 g/mol | C14H4N2Na2O8 | 100 MG
SDP

Catalog No. 5000215870
Encompass_Preferred

Pyrroloquinoline quinone disodium salt is the sodium salt form of PQQ, a redox cofactor used as a research reagent for studies of redox biology and immune function. It is supplied as a pink to red solid with high reported purity and defined storage recommendations.

  • High purity: 98.0%.
  • Molecular weight: 374.17 g/mol.
  • Chemical formula: C14H4N2Na2O8.
  • Appearance: pink to red solid.
  • Storage: store sealed at 4°C, away from moisture.
  • Applications: suitable for redox and immune function research.

Catalog No. 50-002-15870 Supplier Medchemexpress LLC Supplier No. HY100196A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrroloquinoline quinone disodium salt is the sodium salt form of PQQ, a redox cofactor used as a research reagent for studies of redox biology and immune function. It is supplied as a pink to red solid with high reported purity and defined storage recommendations.

  • High purity: 98.0%.
  • Molecular weight: 374.17 g/mol.
  • Chemical formula: C14H4N2Na2O8.
  • Appearance: pink to red solid.
  • Storage: store sealed at 4°C, away from moisture.
  • Applications: suitable for redox and immune function research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.