Learn More
Medchemexpress LLC Pyrroloquinoline quinone disodium salt | 122628-50-6 | 98.0% | 374.17 g/mol | C14H4N2Na2O8 | 50 MG
SDP
Catalog No. 5000215228
Encompass_Preferred
Pyrroloquinoline quinone disodium salt is the disodium salt form of the redox cofactor PQQ supplied for research and analytical applications. The material is a pink to red solid used in studies of metabolism, oxidative stress, and cellular signaling. It is provided as a small powdered reagent with recommended sealed storage to preserve stability.
  • Redox co-factor used in metabolic and oxidative stress research.
  • High purity suitable for analytical and biological applications.
  • Pink to red solid that is easy to weigh and handle.
  • Stable when stored sealed and protected from moisture.
  • Available in small powder pack sizes for laboratory use.
Catalog No. 50-002-15228 Supplier Medchemexpress LLC Supplier No. HY100196A50MG
Pyrroloquinoline quinone disodium salt is the disodium salt form of the redox cofactor PQQ supplied for research and analytical applications. The material is a pink to red solid used in studies of metabolism, oxidative stress, and cellular signaling. It is provided as a small powdered reagent with recommended sealed storage to preserve stability.
  • Redox co-factor used in metabolic and oxidative stress research.
  • High purity suitable for analytical and biological applications.
  • Pink to red solid that is easy to weigh and handle.
  • Stable when stored sealed and protected from moisture.
  • Available in small powder pack sizes for laboratory use.