Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrromethene 556 | 121461-69-6 | 99.0% | 466.20 g·mol⁻1 | C14H15BF2N2Na2O6S2 | 5 MG
SDP

Catalog No. 5000282129
Encompass_Preferred

Pyrromethene 556 is a green-fluorescent pyrromethene dye used as a polar tracer and as a fluorescent gain medium in research. It is employed for studies of membrane fusion, lysis, gap-junctional communication, and liposome or cell-volume assays, offering strong absorption and emission in polar solvents and high fluorescence efficiency.

  • High molar absorptivity at 492 nm in methanol.
  • Emission peak around 533 nm in methanol.
  • High fluorescence quantum yield (~0.73 in water).
  • Solid form with high reported purity (~99.0%).
  • Supplied in small research quantities (5 mg).
  • Soluble in common polar solvents such as methanol, DMF, and water.
  • Suitable for membrane and liposome assays and laser dye applications.

Catalog No. 50-002-82129 Supplier Medchemexpress LLC Supplier No. HYW2481185MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrromethene 556 is a green-fluorescent pyrromethene dye used as a polar tracer and as a fluorescent gain medium in research. It is employed for studies of membrane fusion, lysis, gap-junctional communication, and liposome or cell-volume assays, offering strong absorption and emission in polar solvents and high fluorescence efficiency.

  • High molar absorptivity at 492 nm in methanol.
  • Emission peak around 533 nm in methanol.
  • High fluorescence quantum yield (~0.73 in water).
  • Solid form with high reported purity (~99.0%).
  • Supplied in small research quantities (5 mg).
  • Soluble in common polar solvents such as methanol, DMF, and water.
  • Suitable for membrane and liposome assays and laser dye applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.