Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyrromethene 556 | 121461-69-6 | 99.0% | 466.20 g/mol | C14H15BF2N2Na2O6S2 | 25 MG
SDP

Catalog No. 5000280698
Encompass_Preferred

Pyrromethene 556 (PM556) is a green-fluorescent polar tracer dye used to investigate membrane fusion, lysis, gap-junctional communication, and to detect volume changes in cells or liposomes. Supplied as a high-purity solid, it is suitable for fluorescence assays and membrane biophysics experiments.

  • Green-fluorescent polar tracer dye for membrane and cell studies.
  • High purity: 99.03% (as supplied).
  • Molecular weight 466.20 g/mol; molecular formula C14H15BF2N2Na2O6S2.
  • Soluble in common organic solvents and compatible with fluorescence assays.
  • Used to detect volume changes, membrane fusion, lysis, and gap-junctional communication.

Catalog No. 50-002-80698 Supplier Medchemexpress LLC Supplier No. HYW24811825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrromethene 556 (PM556) is a green-fluorescent polar tracer dye used to investigate membrane fusion, lysis, gap-junctional communication, and to detect volume changes in cells or liposomes. Supplied as a high-purity solid, it is suitable for fluorescence assays and membrane biophysics experiments.

  • Green-fluorescent polar tracer dye for membrane and cell studies.
  • High purity: 99.03% (as supplied).
  • Molecular weight 466.20 g/mol; molecular formula C14H15BF2N2Na2O6S2.
  • Soluble in common organic solvents and compatible with fluorescence assays.
  • Used to detect volume changes, membrane fusion, lysis, and gap-junctional communication.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.