Learn More
Medchemexpress LLC Pyrromethene 556 | 121461-69-6 | MFCD00467373 | 99.0% | 466.20 g/mol | C14H15BF2N2Na2O6S2 | 10 MG
SDP
Catalog No. 5000280855
Encompass_Preferred
Pyrromethene 556 is a green-fluorescent polar tracer dye used to investigate membrane fusion, lysis, gap-junctional communication, and to detect volume changes in cells or liposomes. The compound is supplied as a yellow-orange solid with high purity and defined storage recommendations to preserve fluorescence and chemical stability.
  • Green-fluorescent tracer suitable for membrane and cell volume studies.
  • High purity for reproducible experimental results.
  • Solid, yellow-orange material easy to weigh and dissolve.
  • Multiple package sizes available for experimental flexibility.
  • Stable when stored sealed at 4°C and protected from moisture and light.
  • Recommended solvent storage at -80°C for long term or -20°C for short term.
Catalog No. 50-002-80855 Supplier Medchemexpress LLC Supplier No. HYW24811810MG
Pyrromethene 556 is a green-fluorescent polar tracer dye used to investigate membrane fusion, lysis, gap-junctional communication, and to detect volume changes in cells or liposomes. The compound is supplied as a yellow-orange solid with high purity and defined storage recommendations to preserve fluorescence and chemical stability.
  • Green-fluorescent tracer suitable for membrane and cell volume studies.
  • High purity for reproducible experimental results.
  • Solid, yellow-orange material easy to weigh and dissolve.
  • Multiple package sizes available for experimental flexibility.
  • Stable when stored sealed at 4°C and protected from moisture and light.
  • Recommended solvent storage at -80°C for long term or -20°C for short term.