Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrromethene 580 97.0+%, TCI America™
SDP

Catalog No. B4489200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4489200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4489200MG Supplier TCI America Supplier No. B4489200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 151486-56-5
Molecular Formula C22H33BF2N2
Molecular Weight (g/mol) 374.327
InChI Key XWUUMDJCWNZHAZ-UHFFFAOYSA-N
Synonym Pyrromethene 580
PubChem CID 15294195
SMILES [B-]1(N2C(=C(C(=C2C(=C3[N+]1=C(C(=C3C)CCCC)C)C)C)CCCC)C)(F)F

Specifications

Melting Point 186°C
Quantity 200 mg
Formula Weight 374.33
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [[(4-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](dif
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.