Learn More
Cayman Chemical Pyrrophenon 10mg
SDP
Catalog No. 502955578
Encompass_Preferred
Reversible, selective inhibitor of cPLA2 with an IC50 value of 4.2 nM in enzyme assays; potently blocks the release of AA and the production of PGE2 and LTC4 in cells (IC50 = 24, 25, and 14 nM, respectively)
Catalog No. 50-295-5578 Supplier Cayman Chemical Supplier No. 1329410 MG
Reversible, selective inhibitor of cPLA2 with an IC50 value of 4.2 nM in enzyme assays; potently blocks the release of AA and the production of PGE2 and LTC4 in cells (IC50 = 24, 25, and 14 nM, respectively)